Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

m-PEG16-alcohol 95%

Supplier: AMBEED, INC
Warning

A750778-100MG A750778-250MG
76652-340EA 52 USD InStock
76652-340 76652-342
m-PEG16-alcohol 95%
m-PEG16-alcohol
Precaución: for Research Use Only


Formula: C₃₃H₆₈O₁₇
MDL Number: MFCD27977525
CAS Number: 133604-58-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty