Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium 2,6-dimethoxy-3-pyridinetrifluoroborate 98%

Supplier: AMBEED, INC
Warning

A753267-1G A753267-250MG A753267-5G
76652-454EA 207 USD InStock
76652-454 76652-456 76652-458
Potassium 2,6-dimethoxy-3-pyridinetrifluoroborate 98%
Potassium 2,6-dimethoxy-3-pyridinetrifluoroborate
Precaución: for Research Use Only


Formula: C₇H₈BF₃KNO₂
MDL Number: MFCD22124437
CAS Number: 1374247-42-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty