tert-Butyl 1-bromo-2-oxo-6,9,12-trioxa-3-azapentadecan-15-oate 95%

Supplier: AMBEED, INC
Warning

A753966-100MG A753966-1G A753966-250MG
76652-516EA 5 USD
76652-516 76652-518 76652-520
tert-Butyl 1-bromo-2-oxo-6,9,12-trioxa-3-azapentadecan-15-oate 95%
tert-Butyl 1-bromo-2-oxo-6,9,12-trioxa-3-azapentadecan-15-oate

Precaución: for Research Use Only

Formula: C₁₅H₂₈BrNO₆
MW: 398.2899 g/mol
MDL Number: MFCD28505521
CAS Number: 1807537-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR