Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

16-Bromo-2,5,8,11,14-pentaoxahexadecane 95%

Supplier: AMBEED, INC
Danger

A754002-100MG A754002-1G A754002-250MG A754002-5G
76652-522EA 30 USD InStock
76652-522 76652-524 76652-526 76652-528
16-Bromo-2,5,8,11,14-pentaoxahexadecane 95%
16-Bromo-2,5,8,11,14-pentaoxahexadecane
Precaución: for Research Use Only


Formula: C₁₁H₂₃BrO₅
MDL Number: MFCD28950766
CAS Number: 854601-80-2
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty