Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ald-PEG4-t-butyl ester 95%

Supplier: AMBEED, INC
Warning

A754136-1G A754136-250MG
76652-530EA 89 USD InStock
76652-530 76652-532
Ald-PEG4-t-butyl ester 95%
Ald-PEG4-t-butyl ester
Precaución: for Research Use Only


Formula: C₁₅H₂₈O₇
MDL Number: MFCD25424117
CAS Number: 1415329-20-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty