Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Oxo-1-phenyl-2,7,10,13,16,19,22,25,28-nonaoxa-4-azahentriacontan-31-oic acid 95%

Supplier: AMBEED, INC
Warning

A755570-1G A755570-250MG A755570-5G
76652-566EA 158 USD InStock
76652-566 76652-568 76652-570
3-Oxo-1-phenyl-2,7,10,13,16,19,22,25,28-nonaoxa-4-azahentriacontan-31-oic acid 95%
3-Oxo-1-phenyl-2,7,10,13,16,19,22,25,28-nonaoxa-4-azahentriacontan-31-oic acid
Precaución: for Research Use Only


Formula: C₂₇H₄₅NO₁₂
MDL Number: MFCD11041145
CAS Number: 1334177-87-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty