Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl (2-((2-aminoethyl)disulfanyl)ethyl)carbamate 95%

Supplier: AMBEED, INC
Danger

A756290-5G A756290-250MG A756290-25G
76652-594EA 350 USD InStock
76652-594 77622-818 77622-820
tert-Butyl (2-((2-aminoethyl)disulfanyl)ethyl)carbamate 95%
tert-Butyl (2-((2-aminoethyl)disulfanyl)ethyl)carbamate
Precaución: For research use only.


Formula: C₉H₂₀N₂O₂S₂
MW: 252.3973 g/mol
MDL Number: MFCD19443372
CAS Number: 485800-26-8
UN: 2735
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty