Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-(6-Methyl-1,2,4,5-tetrazin-3-yl)phenyl)acetic acid 95%

Supplier: AMBEED, INC
Warning

A757373-100MG A757373-1G A757373-250MG
76652-618EA 99 USD InStock
76652-618 76652-620 76652-622
2-(4-(6-Methyl-1,2,4,5-tetrazin-3-yl)phenyl)acetic acid 95%
2-(4-(6-Methyl-1,2,4,5-tetrazin-3-yl)phenyl)acetic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₀N₄O₂
MDL Number: MFCD28334556
CAS Number: 1380500-88-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty