Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

O-(3,6,9,12-Tetraoxapentadec-14-yn-1-yl)hydroxylamine 95%

Supplier: AMBEED, INC
Danger

A762213-100MG A762213-1G A762213-250MG
76652-750EA 15 USD InStock
76652-750 76652-752 76652-754
O-(3,6,9,12-Tetraoxapentadec-14-yn-1-yl)hydroxylamine 95%
O-(3,6,9,12-Tetraoxapentadec-14-yn-1-yl)hydroxylamine
Precaución: for Research Use Only


Formula: C₁₁H₂₁NO₅
MDL Number: MFCD29052160
CAS Number: 1835759-78-8
UN: 1325
ADR: 4.1,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty