Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-ACetamido-2,3-dihydro-1h-indene-5-sulfonyl chloride 95%

Supplier: AMBEED, INC
Danger

A762434-100MG A762434-1G A762434-250MG
76652-756EA 59 USD InStock
76652-756 76652-758 76652-760
2-ACetamido-2,3-dihydro-1h-indene-5-sulfonyl chloride 95%
2-ACetamido-2,3-dihydro-1h-indene-5-sulfonyl chloride
Precaución: for Research Use Only


Formula: C₁₁H₁₂ClNO₃S
MDL Number: MFCD27500981
CAS Number: 74124-92-8
UN: 2923
ADR: 8,6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty