Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

CADD522 95%

Supplier: AMBEED, INC
Warning

A773482-10MG A773482-1MG A773482-25MG A773482-50MG A773482-5MG
76652-912EA 37 USD InStock
76652-912 76652-914 76652-916 76652-918 76652-920
CADD522 95%
CADD522
Precaución: for Research Use Only


Formula: C₁₅H₁₃Cl₂NO₃
MW: 326.17 g/mol
MDL Number: MFCD00167693
CAS Number: 199735-88-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty