Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)benzoyl chloride 98%

Supplier: AMBEED, INC
Danger

A777528-1G A777528-250MG A777528-5G
76652-960EA 61 USD InStock
76652-960 76652-962 76652-964
3-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)benzoyl chloride 98%
3-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)benzoyl chloride
Precaución: for Research Use Only


Formula: C₁₂H₁₄BClO₃
MW: 252.50 g/mol
UN: 3261
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty