Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4,6-Trihydroxy-3-methylbenzaldehyde 97%

Supplier: AMBEED, INC
Warning

A778605-100MG A778605-1G
76652-976EA 100 USD InStock
76652-976 76652-978
2,4,6-Trihydroxy-3-methylbenzaldehyde 97%
2,4,6-Trihydroxy-3-methylbenzaldehyde
Precaución: for Research Use Only


Formula: C₈H₈O₄
MDL Number: MFCD24674224
CAS Number: 55743-13-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty