Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2'-[PRopane-2,2-diylbis(thio)]diacetic acid 95%

Supplier: AMBEED, INC
Warning

A780698-100MG A780698-1G A780698-250MG
76653-000EA 99 USD InStock
76653-000 76653-002 77622-930
2,2'-[PRopane-2,2-diylbis(thio)]diacetic acid 95%
2,2'-[PRopane-2,2-diylbis(thio)]diacetic acid
Precaución: for Research Use Only


Formula: C₇H₁₂O₄S₂
MW: 224.2978 g/mol
MDL Number: MFCD01830380
CAS Number: 4265-58-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty