Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 15-bromopentadecanoate 97%

Supplier: AMBEED, INC
Warning

A783597-100MG A783597-1G A783597-250MG A783597-5G A783597-25G
76653-026EA 15 USD InStock
76653-026 76653-028 76653-030 76653-032 77749-548
tert-Butyl 15-bromopentadecanoate 97%
tert-Butyl 15-bromopentadecanoate
Precaución: for Research Use Only


Formula: C₁₉H₃₇BrO₂
MW: 377.3999 g/mol
CAS Number: 905302-64-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty