Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Iridium(1+), [4,4'-bis(1,1-dimethylethyl)-2,2'-bipyridine-κN1,κN1']bis[5-fluoro-2-(5-methyl-2-pyridinyl-κN)phenyl-κC]-, (OC-6-33)-, hexafluorophosphate(1-) (1:1) 97%

Supplier: AMBEED, INC
Warning

A785897-100MG A785897-1G A785897-250MG A785897-50MG
76653-064EA 43 USD InStock
76653-064 76653-066 76653-068 76653-070
Iridium(1+), [4,4'-bis(1,1-dimethylethyl)-2,2'-bipyridine-κN1,κN1']bis[5-fluoro-2-(5-methyl-2-pyridinyl-κN)phenyl-κC]-, (OC-6-33)-, hexafluorophosphate(1-) (1:1) 97%
Iridium(1+), [4,4'-bis(1,1-dimethylethyl)-2,2'-bipyridine-κN1,κN1']bis[5-fluoro-2-(5-methyl-2-pyridinyl-κN)phenyl-κC]-, (OC-6-33)-, hexafluorophosphate(1-) (1:1)
Precaución: for Research Use Only


Formula: C₄₂H₄₂F₈IrN₄P
MDL Number: MFCD31707653
CAS Number: 808142-88-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty