Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(3-Fluoro-4-((methylsulfonyl)methyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%

Supplier: AMBEED, INC
Warning

A788923-100MG A788923-1G A788923-250MG
76653-104EA 37 USD InStock
76653-104 76653-106 76653-108
2-(3-Fluoro-4-((methylsulfonyl)methyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
2-(3-Fluoro-4-((methylsulfonyl)methyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Precaución: for Research Use Only


Formula: C₁₄H₂₀BFO₄S
MW: 314.1806 g/mol
MDL Number: MFCD18760488
CAS Number: 1351499-67-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty