Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 5-nitrothiophene-2-carboxylate 95%

Supplier: AMBEED, INC
Warning

A790389-100MG A790389-1G A790389-250MG A790389-5G
76653-146EA 5 USD InStock
76653-146 76653-148 76653-150 76653-152
tert-Butyl 5-nitrothiophene-2-carboxylate 95%
tert-Butyl 5-nitrothiophene-2-carboxylate
Precaución: for Research Use Only


Formula: C₉H₁₁NO₄S
MW: 229.2529 g/mol
MDL Number: MFCD00159549
CAS Number: 62224-28-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty