Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-tert-Butyl-4-nitro-benzamide 95%

Supplier: AMBEED, INC
Warning

A790734-100G A790734-25G A790734-5G
76653-162EA 89 USD InStock
76653-162 76653-164 76653-166
N-tert-Butyl-4-nitro-benzamide 95%
N-tert-Butyl-4-nitro-benzamide
Precaución: for Research Use Only


Formula: C₁₁H₁₄N₂O₃
MW: 222.2405 g/mol
MDL Number: MFCD00452131
CAS Number: 42498-30-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty