Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-(2-methylpropoxy)benzonitrile 95%

Supplier: AMBEED, INC
Warning

A792618-1G A792618-250MG A792618-5G
76653-240EA 167 USD InStock
76653-240 76653-242 76653-244
4-Bromo-2-(2-methylpropoxy)benzonitrile 95%
4-Bromo-2-(2-methylpropoxy)benzonitrile
Precaución: for Research Use Only


Formula: C₁₁H₁₂BrNO
MW: 254.1231 g/mol
MDL Number: MFCD29043877
CAS Number: 1597288-94-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty