Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-Bromo-5-chlorophenyl)(piperidin-1-yl)methanone 95%

Supplier: AMBEED, INC
Warning

A793078-1G A793078-250MG A793078-5G
76653-266EA 166 USD InStock
76653-266 76653-268 76653-270
(3-Bromo-5-chlorophenyl)(piperidin-1-yl)methanone 95%
(3-Bromo-5-chlorophenyl)(piperidin-1-yl)methanone
Precaución: for Research Use Only


Formula: C₁₂H₁₃BrClNO
MDL Number: MFCD29043944
CAS Number: 1327121-80-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty