Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2,2-Trifluoro-N-(3-hydroxyphenyl)acetamide 95%

Supplier: AMBEED, INC
Warning

A798405-100MG A798405-1G A798405-250MG
76653-376EA 24 USD InStock
76653-376 76653-378 76653-380
2,2,2-Trifluoro-N-(3-hydroxyphenyl)acetamide 95%
2,2,2-Trifluoro-N-(3-hydroxyphenyl)acetamide
Precaución: for Research Use Only


Formula: C₈H₆F₃NO₂
MDL Number: MFCD20366244
CAS Number: 14983-08-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty