Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Diiodophenol 95%

Supplier: AMBEED, INC
Warning

A802550-100MG A802550-1G A802550-250MG
76653-478EA 22 USD InStock
76653-478 76653-480 76653-482
2,4-Diiodophenol 95%
2,4-Diiodophenol
Precaución: for Research Use Only


Formula: C₆H₄I₂O
MDL Number: MFCD01707535
CAS Number: 2012-29-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty