Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-5-fluoro-2-methylbenzoic acid 95%

Supplier: AMBEED, INC
Warning

A802555-100G A802555-1G A802555-25G A802555-5G
76653-488EA 504 USD InStock
76653-488 76653-490 76653-492 76653-494
3-Chloro-5-fluoro-2-methylbenzoic acid 95%
3-Chloro-5-fluoro-2-methylbenzoic acid
Precaución: for Research Use Only


Formula: C₈H₆ClFO₂
MDL Number: MFCD19440350
CAS Number: 1379210-16-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty