Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-5-chloro-6-ethylpyrazine-2-carbonitrile 98%

Supplier: AMBEED, INC
Warning

A802798-1G A802798-250MG A802798-25G A802798-5G
76653-496EA 16 USD InStock
76653-496 76653-498 76653-500 76653-502
3-Bromo-5-chloro-6-ethylpyrazine-2-carbonitrile 98%
3-Bromo-5-chloro-6-ethylpyrazine-2-carbonitrile
Precaución: for Research Use Only


Formula: C₇H₅BrClN₃
MDL Number: MFCD30532902
CAS Number: 2043020-03-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty