Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2-Fluorophenyl)pentan-1-one 95%

Supplier: AMBEED, INC
Danger

A806491-100MG A806491-1G A806491-250MG
76653-548EA 71 USD InStock
76653-548 76653-550 76653-552
1-(2-Fluorophenyl)pentan-1-one 95%
1-(2-Fluorophenyl)pentan-1-one
Precaución: for Research Use Only


Formula: C₁₁H₁₃FO
MDL Number: MFCD14533473
CAS Number: 42916-67-6
UN: 2810
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty