Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-3-methyl-pyridine-2-carboxylic acid methyl ester 96%

Supplier: AMBEED, INC
Warning

A807295-100MG A807295-250MG
76653-582EA 41 USD InStock
76653-582 76653-584
6-Bromo-3-methyl-pyridine-2-carboxylic acid methyl ester 96%
6-Bromo-3-methyl-pyridine-2-carboxylic acid methyl ester
Precaución: for Research Use Only


Formula: C₈H₈BrNO₂
MDL Number: MFCD26517010
CAS Number: 1402666-66-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty