Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Decacarbonyldimanganese 98%

Supplier: AMBEED, INC
Danger

A810679-100MG A810679-250MG
76653-642EA 25 USD InStock
76653-642 76653-644
Decacarbonyldimanganese 98%
Decacarbonyldimanganese
Precaución: for Research Use Only


Formula: C₁₀Mn₂O₁₀
MW: 389.98 g/mol
MDL Number: MFCD00011115
CAS Number: 10170-69-1
UN: 3466
ADR: 6.1 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty