Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Triethylene glycol-2-bromoethyl methyl ether 95%

Supplier: AMBEED, INC
Warning

A814563-1G A814563-250MG A814563-5G
76653-718EA 42 USD InStock
76653-718 76653-720 76653-722
Triethylene glycol-2-bromoethyl methyl ether 95%
Triethylene glycol-2-bromoethyl methyl ether
Precaución: for Research Use Only


Formula: C₉H₁₉BrO₄
MDL Number: MFCD12545963
CAS Number: 110429-45-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty