Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

17-(tert-Butoxy)-17-oxoheptadecanoic acid 97%

Supplier: AMBEED, INC
Warning

A814783-100MG A814783-1G A814783-250MG
76653-728EA 6 USD InStock
76653-728 76653-730 76653-732
17-(tert-Butoxy)-17-oxoheptadecanoic acid 97%
17-(tert-Butoxy)-17-oxoheptadecanoic acid
Precaución: for Research Use Only


Formula: C₂₁H₄₀O₄
MDL Number: MFCD30737822
CAS Number: 905302-44-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty