Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl (4-(1,8-naphthyridin-2-yl)butyl)carbamate 95%

Supplier: AMBEED, INC
Warning

A815604-100MG A815604-1G A815604-250MG
76653-746EA 79 USD InStock
76653-746 76653-748 76653-750
tert-Butyl (4-(1,8-naphthyridin-2-yl)butyl)carbamate 95%
tert-Butyl (4-(1,8-naphthyridin-2-yl)butyl)carbamate
Precaución: for Research Use Only


Formula: C₁₇H₂₃N₃O₂
MDL Number: MFCD31556074
CAS Number: 380382-82-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty