Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2-Hydroxyethyl)-3-methylimidazolium bis(trifluoromethanesulfonyl)imide 97%

Supplier: AMBEED, INC
Warning

A819497-1G A819497-25G A819497-5G
76653-804EA 6 USD InStock
76653-804 76653-806 76653-808
1-(2-Hydroxyethyl)-3-methylimidazolium bis(trifluoromethanesulfonyl)imide 97%
1-(2-Hydroxyethyl)-3-methylimidazolium bis(trifluoromethanesulfonyl)imide
Precaución: for Research Use Only


Formula: C₈H₁₁F₆N₃O₅S₂
MW: 407.30 g/mol
MDL Number: MFCD09038874
CAS Number: 174899-86-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty