Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Butane-1-sulfonyl)phenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A821085-100MG A821085-1G A821085-250MG A821085-5G
76653-820EA 64 USD InStock
76653-820 76653-822 76653-824 76653-826
3-(Butane-1-sulfonyl)phenylboronic acid 98%
3-(Butane-1-sulfonyl)phenylboronic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₅BO₄S
MDL Number: MFCD27935662
CAS Number: 2377611-73-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty