Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-2-Amino-3-(4-((2-nitrobenzyl)oxy)phenyl)propanoic acid 98%

Supplier: AMBEED, INC
Warning

A823394-100MG A823394-1G A823394-250MG
76653-862EA 190 USD InStock
76653-862 76653-864 76653-866
(S)-2-Amino-3-(4-((2-nitrobenzyl)oxy)phenyl)propanoic acid 98%
(S)-2-Amino-3-(4-((2-nitrobenzyl)oxy)phenyl)propanoic acid
Precaución: for Research Use Only


Formula: C₁₆H₁₆N₂O₅
MW: 316.3086 g/mol
MDL Number: MFCD31559511
CAS Number: 184591-51-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty