Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-Arg(Boc)₂-OH (symetrical) 95%

Supplier: AMBEED, INC
Warning

A826210-100MG A826210-250MG A826210-10G
76653-904EA 13 USD InStock
76653-904 77618-318 77749-617
Boc-Arg(Boc)₂-OH (symetrical) 95%
Boc-Arg(Boc)₂-OH (symetrical)
Precaución: for Research Use Only


Formula: C₂₁H₃₈N₄O₈
MDL Number: MFCD00236819
CAS Number: 108787-91-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty