Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

CP-424174 98%

Supplier: AMBEED, INC
Warning

A829204-10MG A829204-1MG A829204-25MG A829204-50MG
76653-956EA 87 USD InStock
76653-956 76653-958 76653-960 76653-962
CP-424174 98%
CP-424174
Precaución: for Research Use Only


Formula: C₂₂H₂₉ClN₂O₄S
MW: 452.99 g/mol
MDL Number: MFCD30537538
CAS Number: 210825-31-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty