Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 6-chloro-5-hydroxynicotinate 95%

Supplier: AMBEED, INC
Warning

A831953-100MG A831953-1G A831953-250MG
76653-998EA 105 USD InStock
76653-998 76654-000 76654-002
Ethyl 6-chloro-5-hydroxynicotinate 95%
Ethyl 6-chloro-5-hydroxynicotinate
Precaución: for Research Use Only


Formula: C₈H₈ClNO₃
MDL Number: MFCD18256611
CAS Number: 1256807-18-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty