Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-2-Methyl-1-pyridin-2-yl-propylamine dihydrochloride 97%

Supplier: AMBEED, INC
Warning

A833744-100MG A833744-1G A833744-250MG
76654-018EA 145 USD InStock
76654-018 76654-020 76654-022
(R)-2-Methyl-1-pyridin-2-yl-propylamine dihydrochloride 97%
(R)-2-Methyl-1-pyridin-2-yl-propylamine dihydrochloride
Precaución: for Research Use Only


Formula: C₉H₁₆Cl₂N₂
MDL Number: MFCD26954804
CAS Number: 1965305-37-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty