Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 1,8-diazaspiro[4.5]decane-8-carboxylate hcl 95%

Supplier: AMBEED, INC
Warning

A835031-100MG A835031-1G A835031-250MG
76654-046EA 17 USD InStock
76654-046 76654-048 76654-050
tert-Butyl 1,8-diazaspiro[4.5]decane-8-carboxylate hcl 95%
tert-Butyl 1,8-diazaspiro[4.5]decane-8-carboxylate hcl
Precaución: for Research Use Only


Formula: C₁₃H₂₅ClN₂O₂
MDL Number: MFCD28892523
CAS Number: 1890715-57-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty