Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 5-(4-methoxy-3-methylphenyl)-3-methyl-1H-pyrrole-2-carboxylate 97%

Supplier: AMBEED, INC
Warning

A838990-100MG A838990-1G A838990-250MG
76654-110EA 13 USD InStock
76654-110 76654-112 76654-114
Ethyl 5-(4-methoxy-3-methylphenyl)-3-methyl-1H-pyrrole-2-carboxylate 97%
Ethyl 5-(4-methoxy-3-methylphenyl)-3-methyl-1H-pyrrole-2-carboxylate
Precaución: for Research Use Only


Formula: C₁₆H₁₉NO₃
MW: 273.3270 g/mol
CAS Number: 2104163-54-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty