Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cyclobutylboronic acid MIDA ester 97%

Supplier: AMBEED, INC
Warning

A841811-1G A841811-250MG A841811-5G
76654-160EA 72 USD InStock
76654-160 76654-162 76654-164
Cyclobutylboronic acid MIDA ester 97%
Cyclobutylboronic acid MIDA ester
Precaución: for Research Use Only


Formula: C₉H₁₄BNO₄
MDL Number: MFCD15144797
CAS Number: 1104637-37-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty