Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(3,5,5-Trimethylcyclohex-2-en-1-ylidene)propanedinitrile 95%

Supplier: AMBEED, INC
Warning

A844213-100MG A844213-1G A844213-250MG A844213-5G
76654-218EA 6 USD InStock
76654-218 76654-220 76654-222 77623-028
2-(3,5,5-Trimethylcyclohex-2-en-1-ylidene)propanedinitrile 95%
2-(3,5,5-Trimethylcyclohex-2-en-1-ylidene)propanedinitrile
Precaución: for Research Use Only


Formula: C₁₂H₁₄N₂
MDL Number: MFCD00228058
CAS Number: 23051-44-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty