Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N4,N4-Dimethylpyridine-2,4-diamine 95%

Supplier: AMBEED, INC
Warning

A846664-1G A846664-250MG
76654-260EA 496 USD InStock
76654-260 76654-262
N4,N4-Dimethylpyridine-2,4-diamine 95%
N4,N4-Dimethylpyridine-2,4-diamine
Precaución: for Research Use Only


Formula: C₇H₁₁N₃
MDL Number: MFCD18384431
CAS Number: 50426-31-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty