Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium oxane-4-sulfinate 98%

Supplier: AMBEED, INC
Warning

A848487-100MG A848487-1G A848487-250MG
76654-294EA 100 USD InStock
76654-294 76654-296 76654-298
Sodium oxane-4-sulfinate 98%
Sodium oxane-4-sulfinate
Precaución: for Research Use Only


Formula: C₅H₉NaO₃S
MDL Number: MFCD24113367
CAS Number: 1516140-99-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty