Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Bromomethyl)quinoline hydrobromide 95%

Supplier: AMBEED, INC
Danger

A851575-100MG A851575-250MG
76654-356EA 120 USD InStock
76654-356 76654-358
3-(Bromomethyl)quinoline hydrobromide 95%
3-(Bromomethyl)quinoline hydrobromide
Precaución: for Research Use Only


Formula: C₁₀H₉Br₂N
MDL Number: MFCD28383872
CAS Number: 1192352-34-3
UN: 3261
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty