Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Guanidine hydrobromide 98% (by titrimetric analysis)

Supplier: AMBEED, INC
Warning

A854196-250MG A854196-5G
76654-408EA 19 USD InStock
76654-408 76654-410
Guanidine hydrobromide 98% (by titrimetric analysis)
Guanidine hydrobromide
Precaución: for Research Use Only


Formula: CH₅N₃
MDL Number: MFCD03094062
CAS Number: 19244-98-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty