Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-3-(1H-indol-3-yl)propan-1-ol 98%

Supplier: AMBEED, INC
Warning

A856374-100MG A856374-1G A856374-250MG A856374-25G A856374-5G
76654-436EA 23 USD InStock
76654-436 76654-438 76654-440 76654-442 77749-663
2-Amino-3-(1H-indol-3-yl)propan-1-ol 98%
2-Amino-3-(1H-indol-3-yl)propan-1-ol
Precaución: for Research Use Only


Formula: C₁₁H₁₄N₂O
MDL Number: MFCD00069710
CAS Number: 526-53-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty