Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 4-(S-methylsulfonimidoyl)piperidine-1-carboxylate 95%

Supplier: AMBEED, INC
Warning

A858424-100MG A858424-250MG
76654-480EA 171 USD InStock
76654-480 76654-482
tert-Butyl 4-(S-methylsulfonimidoyl)piperidine-1-carboxylate 95%
tert-Butyl 4-(S-methylsulfonimidoyl)piperidine-1-carboxylate
Precaución: for Research Use Only


Formula: C₁₁H₂₂N₂O₃S
MDL Number: MFCD29060362
CAS Number: 1934513-55-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty