Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 2-bromo-4-nitrobenzoate 95%

Supplier: AMBEED, INC
Warning

A863945-1G A863945-250MG A863945-25G
76654-588EA 16 USD InStock
76654-588 76654-590 76654-592
Ethyl 2-bromo-4-nitrobenzoate 95%
Ethyl 2-bromo-4-nitrobenzoate
Precaución: for Research Use Only


Formula: C₉H₈BrNO₄
MDL Number: MFCD12406858
CAS Number: 128566-93-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty