Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl N-{2-[2-(methylamino)ethoxy]ethyl}carbamate 95%

Supplier: AMBEED, INC
Danger

A865968-5G A865968-100MG A865968-250MG
76654-628EA 262 USD InStock
76654-628 77623-072 77623-074
tert-Butyl N-{2-[2-(methylamino)ethoxy]ethyl}carbamate 95%
tert-Butyl N-{2-[2-(methylamino)ethoxy]ethyl}carbamate
Precaución: For research use only.


Formula: C₁₀H₂₂N₂O₃
MDL Number: MFCD25368799
CAS Number: 1185772-28-4
UN: 2735
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty